Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20393

Sigma-Aldrich

Diethylammonium chloride

for synthesis

Synonym(s):

Diethylammonium chloride, Diethylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$43.90
250 G
$54.10
1 KG
$120.00
25 KG
$812.00

$43.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$43.90
250 G
$54.10
1 KG
$120.00
25 KG
$812.00

About This Item

Linear Formula:
(C2H5)2NH2Cl
CAS Number:
Molecular Weight:
109.60
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-541-9
NACRES:
NA.22

$43.90


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.00001 hPa ( 20 °C)

Quality Level

form

crystals

potency

9900 mg/kg LD50, oral (Rat)

pH

4.5-6.5 (20 °C, 10 g/L in H2O)

mp

231.8 °C

solubility

soluble 510 g/L

density

1.07 g/cm3 at 20.1 °C

bulk density

1100 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[N+H2](CC)CC

InChI

1S/C4H11N.ClH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1H

InChI key

HDITUCONWLWUJR-UHFFFAOYSA-N

Application

  • Hygroscopicity of secondary marine organic aerosols: Mixtures of alkylammonium salts and inorganic components.: This study explores the hygroscopic properties of secondary marine organic aerosols, highlighting the role of alkylammonium salts, including diethylammonium chloride, in influencing aerosol behavior in the atmosphere (Asadzadeh et al., 2021).
  • Simulation of Deep Eutectic Solvents′ Interaction with Membranes of Cancer Cells Using COSMO-RS.: This paper discusses the simulation of interactions between deep eutectic solvents containing diethylammonium chloride and cancer cell membranes, providing insights into their potential use in biomedical applications (Mbous et al., 2020).

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service