Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20293

Sigma-Aldrich

Sodium chloroacetate

for synthesis

Synonym(s):

Sodium chloroacetate, Chloroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$57.30
500 G
$76.60
50 KG
$1,440.00

$57.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$57.30
500 G
$76.60
50 KG
$1,440.00

About This Item

Empirical Formula (Hill Notation):
C2H2ClNaO2
CAS Number:
Molecular Weight:
116.48
MDL number:
UNSPSC Code:
12352107
EC Index Number:
223-498-3
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
powder

$57.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

form

powder

potency

95 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

4.5-9.0 (20 °C, 50 g/L in H2O)

transition temp

flash point 270 °C

bulk density

850 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].ClCC(=O)[O-]

InChI

1S/C2H3ClO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

FDRCDNZGSXJAFP-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

518.0 °F

Flash Point(C)

270 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service