Skip to Content
MilliporeSigma
All Photos(1)

Documents

82023

SAFC

Cholesterol Supertrate, Bovine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32011401

biological source

bovine serum

Quality Level

type

for cell culture

shelf life

12 mo.

storage temp.

2-8°C

Storage and Stability

refrigerated

Disclaimer

For research and further manufacturing use only. It is not for use in humans.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service