Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.20162

Sigma-Aldrich

4,4′-Bis(dimethylamino)thiobenzophenone

for synthesis

Synonym(s):

4,4′-Bis(dimethylamino)thiobenzophenone, N,N,N′,N′-Tetramethyl-4,4′-diaminothiobenzophenone, Michler thioketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NC6H4CSC6H4-4-[N(CH3)2]
CAS Number:
Molecular Weight:
284.42
MDL number:
UNSPSC Code:
12352115
EC Index Number:
214-962-6

Assay

≥84.0% (HPLC)

form

powder

mp

198-206 °C

storage temp.

2-30°C

InChI

1S/C17H20N2S/c1-18(2)15-9-5-13(6-10-15)17(20)14-7-11-16(12-8-14)19(3)4/h5-12H,1-4H3

InChI key

KFUJUTFTRXYQMG-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 84.0 % (a/a)
Identity (NMR-spectrum): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service