Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.20022

Sigma-Aldrich

O-Ethylbenzoin

for synthesis

Synonym(s):

O-Ethylbenzoin, Benzoin ethyl ether, 2-Ethoxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO(C2H5)COC6H5
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352115
EC Index Number:
209-366-8
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

mp

59-61 °C (External MSDS)

storage temp.

2-30°C

SMILES string

O(CC)C(c2ccccc2)C(=O)c1ccccc1

InChI

1S/C16H16O2/c1-2-18-16(14-11-7-4-8-12-14)15(17)13-9-5-3-6-10-13/h3-12,16H,2H2,1H3

InChI key

KMNCBSZOIQAUFX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 59 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service