Skip to Content
MilliporeSigma
All Photos(2)

Documents

8.19100

Sigma-Aldrich

2,2-Dichloropropionic acid sodium salt

for synthesis

Synonym(s):

2,2-Dichloropropionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3Cl2NaO2
CAS Number:
Molecular Weight:
164.95
UNSPSC Code:
12352107
EC Index Number:
204-828-5

Assay

≥90.0% (GC)

form

crystals

potency

3860 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

5.5 (20 °C, 100 g/L in H2O)

mp

166.5 °C

solubility

450 g/L

storage temp.

2-30°C

InChI

1S/C3H4Cl2O2.Na/c1-3(4,5)2(6)7;/h1H3,(H,6,7);/q;+1/p-1

InChI key

PDEFQWNXOUGDJR-UHFFFAOYSA-M

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Assay (HClO₄): ≥ 90.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service