Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.19007

Sigma-Aldrich

TOLUENE-4-SULFONIC ACID

65% SOLUTION IN WATER) FOR SYNTHESIS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107

Quality Level

storage temp.

10-30°C

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service