Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18882

Sigma-Aldrich

Chlorosulfonyl isocyanate

for synthesis

Synonym(s):

Chlorosulfonyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CClNO3S
CAS Number:
Molecular Weight:
141.53
MDL number:
UNSPSC Code:
12352118
EC Index Number:
214-715-2

vapor pressure

25.6 hPa ( 20 °C)

Quality Level

form

liquid

potency

640 mg/kg LD50, oral (Rat)

bp

106 °C/1013 hPa

mp

-44 °C

transition temp

flash point >110 °C

density

1.63 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/CClNO3S/c2-7(5,6)3-1-4

InChI key

WRJWRGBVPUUDLA-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.631 - 1.634
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service