Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18732

Sigma-Aldrich

2,3-Hexanedione

for synthesis

Synonym(s):

2,3-Hexanedione, propyl diketone, methylpropylglyoxal

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$120.00
100 ML
$334.00

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$120.00
100 ML
$334.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352115
EC Index Number:
223-350-8
NACRES:
NA.22

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

13 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

245 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-5.9 % (v/v)

bp

130 °C/1013 hPa

mp

-30 °C

transition temp

flash point 28 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C(CCC)C(=O)C

InChI

1S/C6H10O2/c1-3-4-6(8)5(2)7/h3-4H2,1-2H3

InChI key

MWVFCEVNXHTDNF-UHFFFAOYSA-N

Application


  • Synthesis of alpha-hydroxy ketones and vicinal (R, R)-diols by Bacillus clausii DSM 8716 T butanediol dehydrogenase: This study explores the enzymatic reduction of 2,3-hexanedione to produce hydroxy ketones, providing insights into biocatalytic applications (Muschallik et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.934 - 0.940
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT RE 2 Inhalation

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service