Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18639

Sigma-Aldrich

N-Methylbutylamine

for synthesis

Synonym(s):

N-Methylbutylamine, N-Butylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NHCH3
CAS Number:
Molecular Weight:
87.16
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-791-2
Pricing and availability is not currently available.

Assay

≥98.0% (GC)

form

liquid

potency

420 mg/kg LD50, oral (Rat)
932 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3 % (v/v)

bp

90-92 °C/1013 hPa

mp

-75 °C

transition temp

flash point -4 °C

density

0.74 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H13N/c1-3-4-5-6-2/h6H,3-5H2,1-2H3/p+1

InChI key

QCOGKXLOEWLIDC-UHFFFAOYSA-O

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.734 - 0.736
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

-1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service