Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.18567

Sigma-Aldrich

(R)-(+)-1,2-Propylene oxide

for synthesis

Synonym(s):

(R)-(+)-1,2-Propylene oxide, (R)-(+)-1,2-Epoxypropane, (R)-(+)-Methyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
MDL number:
UNSPSC Code:
12352005
EC Index Number:
200-879-2

vapor pressure

588 hPa ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

430 °C (DIN 51794)

potency

380 mg/kg LD50, oral (Rat)
1243 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-37 % (v/v)

bp

34-35 °C/1013 hPa

mp

-112 °C

transition temp

flash point -37 °C

density

0.829 g/cm3 at 20 °C

storage temp.

2-25°C

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/t3-/m1/s1

InChI key

GOOHAUXETOMSMM-GSVOUGTGSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.829 - 0.832
Spec. rotation [α²0/D (undiluted): +13.0 - +15.0 °
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - Pensky-Martens closed cup

Flash Point(C)

-40 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service