Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18214

Sigma-Aldrich

1,3-Dimethyltetrahydro-2(1H)-pyrimidinone

for synthesis

Synonym(s):

1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, Dimethylpropyleneurea, DMPU

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$112.00
500 ML
$290.00
25 KG
$5,400.00

$112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$112.00
500 ML
$290.00
25 KG
$5,400.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352115
EC Index Number:
230-625-6
NACRES:
NA.22

$112.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.5 hPa ( 57 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

1770 mg/kg LD50, oral (Rat)

expl. lim.

1.6-9.3 % (v/v)

pH

11 (20 °C, 100 g/L in H2O)

bp

247 °C/1013 hPa

mp

-24 °C

transition temp

flash point 121 °C

density

1.064 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(CCCN(C1=O)C)C

InChI

1S/C6H12N2O/c1-7-4-3-5-8(2)6(7)9/h3-5H2,1-2H3

InChI key

GUVUOGQBMYCBQP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.062 - 1.066
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service