Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18151

Sigma-Aldrich

Cyclohexyl isocyanide

for synthesis

Synonym(s):

Cyclohexyl isocyanide, Cyclohexyl isonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N
CAS Number:
Molecular Weight:
109.17
MDL number:
UNSPSC Code:
12352117
EC Index Number:
213-238-7
NACRES:
NA.22
Assay:
≥97.0% (GC)

Quality Level

Assay

≥97.0% (GC)

bp

173-176 °C/1013 hPa

transition temp

flash point 77 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[N](C1CCCCC1)[C]

InChI

1S/C7H11N/c1-8-7-5-3-2-4-6-7/h7H,2-6H2

InChI key

XYZMOVWWVXBHDP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.892 - 0.894
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service