Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18097

Sigma-Aldrich

Aluminium triethylate

for synthesis

Synonym(s):

Aluminium triethylate, Aluminium triethoxide, Aluminium triethanolate, Aluminiumethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OC2H5)3
CAS Number:
Molecular Weight:
162.16
MDL number:
UNSPSC Code:
12352103
EC Index Number:
209-105-8

form

pieces

mp

128-131 °C

density

1.142 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/3C2H5O.Al/c3*1-2-3;/h3*2H2,1H3;/q3*-1;+3

InChI key

JPUHCPXFQIXLMW-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 97.0 %
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service