Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.16007

Sigma-Aldrich

(S)-(-)-trans-4-Hydroxyproline

for synthesis

Synonym(s):

(S)-(-)-trans-4-Hydroxyproline, L(-)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
UNSPSC Code:
12352209
EC Index Number:
200-091-9

pH

5.5-6.5 (20 °C, 50 g/L in H2O)

mp

274 °C

solubility

500 g/L

bulk density

610 kg/m3

storage temp.

2-30°C

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)

InChI key

PMMYEEVYMWASQN-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=4 in water): -77.0 - -74.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service