Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

8.16001

Sigma-Aldrich

(S)-(+)-Arginine hydrochloride

for synthesis

Synonym(s):

L-Arginine monohydrochloride, α-Amino-δ-guanidino valerianic acid hydrochloride, alpha-Amino-delta-guanidino valerianic acid hydrochloride, Arg, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$81.10

$81.10


Please contact Customer Service for Availability


Select a Size

Change View
100 G
$81.10

About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
MDL number:
UNSPSC Code:
12352209
EC Index Number:
214-275-1
NACRES:
NA.22

$81.10


Please contact Customer Service for Availability

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<1 Pa ( 20 °C)

Assay

≥99% (silver nitrate titration)

form

solid

potency

12000 mg/kg LD50, oral (Rat)

pH

5.7 (20 °C, 100 g/L in H2O)

mp

>235 °C

solubility

730 g/L

density

1.42 g/cm3 at 20 °C

bulk density

1250 kg/m3

storage temp.

2-30°C

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Analysis Note

Assay (argentometric): ≥ 99.0 %
Spec. rotation [α²0/D (c=8 in HCl 6 mol/l): +21.5 - +23.0 °
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service