Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14925

Sigma-Aldrich

Kryptofix® 222

special quality for synthesis

Synonym(s):

Kryptofix® 222, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N2O6
CAS Number:
Molecular Weight:
376.49
MDL number:
UNSPSC Code:
12352005
EC Index Number:
245-962-4
NACRES:
NA.22

Quality Level

potency

300-2000 mg/kg LD50, oral (Rat)

mp

70-73 °C

storage temp.

2-8°C

InChI

1S/C18H36N2O6/c1-7-21-13-14-24-10-4-20-5-11-25-17-15-22-8-2-19(1)3-9-23-16-18-26-12-6-20/h1-18H2

InChI key

AUFVJZSDSXXFOI-UHFFFAOYSA-N

Application

  • Improved, one-pot synthesis of 6-[(18) F]fluorodopamine and quality control testing for use in patients with neuroblastoma.: This study presents an improved method for synthesizing 6-[(18) F]fluorodopamine using Kryptofix 222. The one-pot synthesis process enhances efficiency and quality control, making it suitable for clinical applications in neuroblastoma patients (Vāvere et al., 2018).
  • Efficient radiosynthesis and non-clinical safety tests of the TSPO radioprobe [(18)F]FEDAC: Prerequisites for clinical application.: Kryptofix 222 is utilized in the radiosynthesis of [(18)F]FEDAC, a radioprobe for TSPO imaging. The study highlights the efficiency and safety of the synthesis process, facilitated by Kryptofix 222, and its potential for clinical application (Kawamura et al., 2016).
  • Fully automated one-pot radiosynthesis of O-(2-[18F]fluoroethyl)-L-tyrosine on the TracerLab FX(FN) module.: This research demonstrates the automated synthesis of O-(2-[18F]fluoroethyl)-L-tyrosine, employing Kryptofix 222 as a phase transfer catalyst. The automated process significantly improves the synthesis efficiency (Bourdier et al., 2011).
  • Electrochemical concentration of no-carrier-added [(18)F]fluoride from [(18)O]water in a disposable microfluidic cell for radiosynthesis of (18)F-labeled radiopharmaceuticals.: Kryptofix 222 plays a crucial role in the electrochemical concentration of [(18)F]fluoride, facilitating the radiosynthesis of various (18)F-labeled radiopharmaceuticals in a microfluidic cell, enhancing the overall efficiency and yield (Saiki et al., 2010).

Analysis Note

Assay (perchloric acid titration): ≥ 99
Purity (GC): ≥ 99
Identity (IR-spectrum): passes test
Melting point: 69 - 72
Biggest singly impurity (GC): ≤ 0.1 %
Acetone (HS-GC): ≤ 5000 ppm
Acetonitrile (HS-GC): ≤ 410 ppm
Dichloromethane (HS-GC): ≤ 600 ppm
n-Hexane (HS-GC): ≤ 290 ppm
Methanol (HS-GC): ≤ 3000 ppm
petroleum ether (HS-GC): ≤ 290 ppm
Pyridine (HS-GC): ≤ 200 ppm
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): ≤ 0.5
E.coli: passes test
Salmonella species: passes test
Staphylococcus aureus: passes test
Pseudomonas aeruginosa: passes test
Colony count (aerobic bacteria): ≤ 5000 CFU/g
Colony count (Yeasts and moulds): ≤ 500 CFU/g
Endotoxins: ≤ 50 I.U./g

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service