Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14894

Sigma-Aldrich

Polyvinyl alcohol, fully hydrolyzed

(Mw approx. 145000) for synthesis

Synonym(s):

Poly(vinyl alcohol), PVA, PVOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
13111042
NACRES:
NA.22

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

mp

160-240 °C

bulk density

400‑670 kg/m3

storage temp.

2-30°C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Controlled Drug Release: Polyvinyl alcohol-based polymeric nanofibers, produced via electrospinning, demonstrated effective controlled release mechanisms for lysozyme, suggesting potential applications in targeted drug delivery systems (Ogawa et al., 2024).
  • Recyclable Hydrogels: Fully hydrolyzed polyvinyl alcohol has been utilized to fabricate recyclable hydrogels at room temperature, highlighting its utility in developing environmentally friendly materials for diverse applications (Liu et al., 2023).
  • Antibacterial Polymeric Films: The hydrolysis degree of polyvinyl alcohol significantly affects the properties of antibacterial polymeric films, which incorporate zinc sulphate for enhanced biomedical applications (Sedlarik et al., 2010).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service