Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14807

Sigma-Aldrich

1,13-Diamino-4,7,10-trioxatridecane

for synthesis

Synonym(s):

1,13-Diamino-4,7,10-trioxatridecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2O3
CAS Number:
Molecular Weight:
220.31
MDL number:
UNSPSC Code:
12352116
EC Index Number:
224-207-2
NACRES:
NA.22

vapor pressure

<0.001 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

260 °C

potency

3160 mg/kg LD50, oral (Rat)
2500 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-4.5 % (v/v)

pH

>12 (20 °C, 100 g/L in H2O)

bp

146-148 °C/1 hPa

mp

-32 °C

transition temp

flash point 139 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H24N2O3/c11-3-1-5-13-7-9-15-10-8-14-6-2-4-12/h1-12H2/p+2

InChI key

JCEZOHLWDIONSP-UHFFFAOYSA-P

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.003 - 1.008
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

282.2 °F

Flash Point(C)

139 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service