Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14763

Sigma-Aldrich

Lithium aluminium hydride

(powder) for synthesis

Synonym(s):

Lithium aluminium hydride, LAH, Lithium alanate, Aluminium lithium hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4AlLi
CAS Number:
Molecular Weight:
37.95
MDL number:
UNSPSC Code:
12352306
EC Index Number:
240-877-9

Quality Level

form

powder

reaction suitability

reagent type: reductant

mp

125 °C (decomposition)

density

0.917 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

2-30°C

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Analysis Note

Assay (by gasvolumetry): ≥ 97.0 %

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service