Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14739

Sigma-Aldrich

N-Methylpiperidine-2-ethanol

for synthesis

Synonym(s):

N-Methylpiperidine-2-ethanol, 2-(2-Hydroxyethyl) 1-methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO
CAS Number:
Molecular Weight:
143.23
UNSPSC Code:
12352104
EC Index Number:
208-554-7

Quality Level

Assay

≥95.0% (GC)

potency

1800 mg/kg LD50, oral (Rat)

refractive index

n20/D 1.48

pH

11.2 (20 °C, 50 g/L in H2O)

transition temp

flash point 106 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H17NO/c1-9-6-3-2-4-8(9)5-7-10/h8,10H,2-7H2,1H3

InChI key

OVMRRCXDBKEQIU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.972 - 0.977
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

222.8 °F

Flash Point(C)

106 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service