Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14675

Sigma-Aldrich

2-Hydroxy-3-methyl-2-cyclopentene-1-one

for synthesis

Synonym(s):

2-Hydroxy-3-methyl-2-cyclopentene-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
UNSPSC Code:
12352115
EC Index Number:
201-303-2
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

crystals

mp

104-107 °C

transition temp

flash point 100 °C

storage temp.

2-8°C

InChI

1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h8H,2-3H2,1H3

InChI key

CFAKWWQIUFSQFU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 104 °C
Melting range (upper value): ≤ 107 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service