Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14648

Sigma-Aldrich

4-Aminodiphenylamine

for synthesis

Synonym(s):

4-Aminodiphenylamine, Diphenyl black, N-Phenyl-p-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)NH(C6H4)NH2
CAS Number:
Molecular Weight:
184.24
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-951-9

vapor pressure

1 hPa

form

solid

autoignition temp.

>500 °C

potency

464 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

8.9 (20 °C, 0.5 g/L in H2O)

bp

354 °C/1013 hPa

mp

72-75 °C

transition temp

flash point 199 °C

density

1.09 g/m3

storage temp.

2-30°C

InChI

1S/C12H12N2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,14H,13H2

InChI key

ATGUVEKSASEFFO-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 72 °C
Melting range (upper value): ≤ 75 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

199 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service