Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14388

Sigma-Aldrich

(1S)-(-)-3-Oxocamphorsulfonylimine

for synthesis

Synonym(s):

(1S)-(-)-3-Oxocamphorsulfonylimine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
227.28
UNSPSC Code:
12352115
NACRES:
NA.22

Quality Level

form

powder

mp

195-198 °C

storage temp.

2-30°C

General description

for synthesis

Application

(1S)-(-)-3-Oxocamphorsulfonylimine for synthesis. CAS 119106-38-6, molar mass 227.28 g/mol.

Analysis Note

Assay (of S)≥ 98.0 %Melting range (lower value)≥ 195 °CMelting range (upper value)≤ 198 °CSpec. rotation [α²0/D (c=2 in aceton)-192.0 - -190.0 °Identity (IR)passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service