Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.14363

Sigma-Aldrich

N-Alkyl-N,N-dimethyl-N-benzylammonium chloride

for synthesis

Synonym(s):

N-Alkyl-N,N-dimethyl-N-benzylammonium chloride, Benzyldimethylmyristylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352107
EC Index Number:
287-089-1
NACRES:
NA.22

Quality Level

autoignition temp.

>250 °C (Dust)

pH

5-8 (20 °C, 10 g/L in H2O)

mp

73 °C

transition temp

flash point >100 °C

solubility

250 g/L

bulk density

350 kg/m3

storage temp.

2-30°C

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (upper value): ≤ 73 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service