Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14204

Sigma-Aldrich

5,5-Dimethyl-2-nitromethyl-1,3-dioxane

for synthesis

Synonym(s):

5,5-Dimethyl-2-nitromethyl-1,3-dioxane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352102
NACRES:
NA.22

Quality Level

form

liquid

bp

67-69 °C/1 hPa

density

1.14 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])CC1OCC(CO1)(C)C

InChI

1S/C7H13NO4/c1-7(2)4-11-6(12-5-7)3-8(9)10/h6H,3-5H2,1-2H3

InChI key

AVQLWXDZRAOBJX-UHFFFAOYSA-N

General description

for synthesis

Application

5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis. CAS 33884-29-6, molar mass 175.18 g/mol.

Analysis Note

Assay (GC, area%)≥ 95.0 % (a/a)Density (d 20 °C/ 4 °C)1.135 - 1.140Identity (IR)passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service