Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14095

Sigma-Aldrich

Sodium amide crystalline

for synthesis

Synonym(s):

Sodium amide crystalline

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$97.30
1 KG
$425.00

$97.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$97.30
1 KG
$425.00

About This Item

Empirical Formula (Hill Notation):
H2NNa
CAS Number:
Molecular Weight:
39.01
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-971-0
NACRES:
NA.22

$97.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

Assay

≥95.0% (acidimetric)

form

crystals

autoignition temp.

450 °C

bp

400 °C/1013 hPa

mp

210 °C

density

1.39 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Na]N

InChI

1S/H2N.Na/h1H2;/q-1;+1

InChI key

ODZPKZBBUMBTMG-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 95.0 %

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service