8.14079
Iodotrimethylsilane
(stabilised) for synthesis
Synonym(s):
Iodotrimethylsilane, Trimethyliodosilane, Trimethylsilyl iodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
200.1
MDL number:
UNSPSC Code:
12352103
EC Index Number:
240-171-0
Quality Level
transition temp
flash point -31 °C
density
1.47 g/cm3 at 20 °C
InChI
1S/C3H9ISi/c1-5(2,3)4/h1-3H3
InChI key
CSRZQMIRAZTJOY-UHFFFAOYSA-N
General description
(stabilised) for synthesis
Application
Iodotrimethylsilane (stabilised) for synthesis. CAS 16029-98-4, EC Number 240-171-0, chemical formula (CH₃)₃SiI.
Analysis Note
Assay (argentometric)≥ 95.0 %Identity (IR)passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-23.8 °F - closed cup
Flash Point(C)
-31 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service