Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

811771

Sigma-Aldrich

Probumin® Bovine Serum Albumin Polymer Enhanced Euro, 30% Solution

29-31 g/dL protein (Biuret), USA origin, Tested for IgG, Blood banking: suitable, Serology: suitable, enzyme immunoassay: suitable

Sign Into View Organizational & Contract Pricing

Select a Size

10 L
$6,120.00

$6,120.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 L
$6,120.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801

$6,120.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Probumin® Bovine Serum Albumin Polymer Enhanced Euro, 30% Solution, pack of 10 L

description

0.1% sodium azide as preservtive
Contains sodium azide less than 0.1 g/dL

Quality Level

Assay

29-31 g/dL protein basis (biuret)

form

solution

shelf life

36 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pack of 10 L

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL protein (Biuret)

technique(s)

blood typing: suitable

dilution

(Immunohematology)
(Serology)

impurities

Tested for IgG

pH

7.2-7.4

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service