Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.10261

Sigma-Aldrich

Diisoamyl ether

(stabilized) for synthesis

Synonym(s):

Diisoamyl ether, Diisopentyl ether, Isoamyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O
CAS Number:
Molecular Weight:
158.28
MDL number:
UNSPSC Code:
12352112
EC Index Number:
208-857-4
Pricing and availability is not currently available.

vapor pressure

2.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

172-173 °C/1013 hPa

mp

<-75 °C

transition temp

flash point 46 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H22O/c1-9(2)5-7-11-8-6-10(3)4/h9-10H,5-8H2,1-4H3

InChI key

AQZGPSLYZOOYQP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.775 - 0.777
Water (K. F.): ≤ 0.50 %
Peroxide (as H₂O₂): ≤ 0.02 %
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service