Skip to Content
MilliporeSigma
All Photos(1)

Documents

810017

Sigma-Aldrich

Probumin® Bovine Serum Albumin Crystallized Grade

≥99%, USA Origin, Protein standard: suitable, ligand binding assay: suitable, EIA: Suitable ELISA: suitable

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41141801

product name

Probumin® Bovine Serum Albumin Crystallized Grade, pack of 5 kg

biological source

bovine serum

Quality Level

Assay

≥99% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

apparent mol wt 66 kDa

purified by

heat shock fractionation

packaging

pack of 5 kg

manufacturer/tradename

Probumin®

origin

USA origin

technique(s)

ELISA: suitable
enzyme immunoassay: suitable
ligand binding assay: suitable
protein quantification: suitable

impurities

Tested for IgG

loss

≤5%

pH range

6.8-7.2

anion traces

chloride (Cl-): 0.0-3.0 mg/g

cation traces

Fe: 0.0-15.0 μg/g
Na: 0.0-10.0 mg/g

foreign activity

Bluetongue, none detected
Vesicular Stomatitis Virus, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This is a highly purified BSA. One of the highest purity bovine albumins commercially available, minimizing interfering components in sensitive applications. Crystal like presentation of this BSA simplifies handling, weighing, and dispensing. This bovine albumin has a purity specification of >=99%; typical purity is 100%. This BSA may be used for molecular biology applications.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Probumin® BSA grades are distinguished by differences in production. The use of terms such as ′Diagnostic Grade′ or ′Vaccine Grade′ in product names solely indicates that said products are commonly used by customers in those applications. We have not qualified every single product in every conceivable application. Users should qualify Probumin® BSA grades using the specific reagents, conditions, and constraints of their assay.
For further manufacturing use only. Not for human or animal consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service