Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.08653

Sigma-Aldrich

Triphenylarsine

for synthesis

Synonym(s):

Triphenylarsine, Arsenic triphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3As
CAS Number:
Molecular Weight:
306.23
MDL number:
UNSPSC Code:
12352103
EC Index Number:
210-032-9
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

>360 °C/1013 hPa (decomposition)

mp

59-61 °C

transition temp

flash point 265 °C

density

1.22 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C18H15As/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

BPLUKJNHPBNVQL-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 57 °C
Melting range (upper value): ≤ 61 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service