Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.08391

Sigma-Aldrich

Trichloroacetyl chloride

for synthesis

Synonym(s):

Trichloroacetyl chloride, Trichloroacetic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
181.83
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-926-7
NACRES:
NA.22

vapor pressure

23 hPa ( 20 °C)

Quality Level

form

liquid

potency

600 mg/kg LD50, oral (Rat)

pH

1.11 (25 °C, 16.2 g/L in H2O)

kinematic viscosity

13.73 cSt(35 °C)

bp

132 °C/1013 hPa (decomposition)

mp

-57 °C

transition temp

flash point 100 °C

density

1.62 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2Cl4O/c3-1(7)2(4,5)6

InChI key

PVFOMCVHYWHZJE-UHFFFAOYSA-N

General description

for synthesis

Application

Trichloroacetyl chloride for synthesis. CAS 76-02-8, EC Number 200-926-7, chemical formula CCl₃COCl.

Analysis Note

Assay (morpholine method)≥ 96.0 %Density (d 20 °C/ 4 °C)1.616 - 1.621Identity (IR)passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F - open cup

Flash Point(C)

101 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service