Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07469

Sigma-Aldrich

4-Pyridinecarbaldehyde

for synthesis

Synonym(s):

4-Pyridinecarbaldehyde, Pyridine-4-aldehyde, Isonicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO
CAS Number:
Molecular Weight:
107.11
MDL number:
UNSPSC Code:
12352114
EC Index Number:
212-832-3

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

potency

2355 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

7-8 (20 °C in H2O, saturated aqueous solution)

bp

198 °C/1013 hPa

mp

-4 °C

transition temp

flash point 77.8 °C

density

1.138 g/cm3 at 20 °C

InChI

1S/C6H5NO/c8-5-6-1-3-7-4-2-6/h1-5H

InChI key

BGUWFUQJCDRPTL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.138 - 1.142
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

172.0 °F

Flash Point(C)

77.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service