Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07110

Sigma-Aldrich

Palladium(II) chloride

59% Pd, anhydrous, for synthesis

Synonym(s):

Palladium(II) chloride (59% Pd), Palladium dichloride, Hydrochloric acid palladium salt, Palladium Chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$167.00
1 G
$238.00
5 G
$1,660.00
10 G
$3,030.00
50 G
$8,220.00

$167.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$167.00
1 G
$238.00
5 G
$1,660.00
10 G
$3,030.00
50 G
$8,220.00

About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-596-2
NACRES:
NA.22

$167.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Palladium(II) chloride (59% Pd), anhydrous, for synthesis

Quality Level

form

powder

potency

2704 mg/kg LD50, oral (Rat)

pH

2.15 (20 °C, 30 g/L in H2O)

solubility

insoluble 55.6 g/L

density

4.0 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Pd+2].[Cl-].[Cl-]

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Application


  • Determination of free and reversibly-bound sulfite in selected foods by high-performance liquid chromatography with fluorometric detection: This study outlines a method for determining sulfite levels in food, which has implications for understanding the environmental and health impacts of sulfites, including those related to formaldehyde-sodium bisulfite adducts (Chan et al., 2008).



Analysis Note

Assay (ICP-OES): ≥ 99.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service