Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.07044

Sigma-Aldrich

Phenylacetylene

for synthesis

Synonym(s):

Phenylacetylene, Ethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
208-645-1
NACRES:
NA.22

vapor pressure

2.7 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

142-144 °C/1013 hPa

mp

-44.8 °C

transition temp

flash point 27 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7H

InChI key

UEXCJVNBTNXOEH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.929 - 0.934
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service