Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.06116

Sigma-Aldrich

Methyltriphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352101

reaction suitability

reaction type: C-C Bond Formation

Quality Level

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

464.0 °F

Flash Point(C)

> 240 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service