Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.06086

Sigma-Aldrich

α-Methylstyrene

(stabilised with 4-tert-butylpyrocatechol) for synthesis

Synonym(s):

α-Methylstyrene, 2-Phenyl-1-propene, Isopropenylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$73.90

$73.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$73.90

About This Item

Linear Formula:
C6H5C(CH3)=CH2
CAS Number:
Molecular Weight:
118.18
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-705-0
NACRES:
NA.22

$73.90


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

2.9 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

445 °C

potency

4900 mg/kg LD50, oral (Rat)

expl. lim.

0.9-6.6 % (v/v)

pH

5-6 (500 g/L in H2O)

bp

164 °C/1013 hPa

mp

-23 °C

transition temp

flash point 40 °C (DIN 51758)

solubility

560 mg/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

CC(=C)c1ccccc1

InChI

1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

XYLMUPLGERFSHI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.908 - 0.910
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service