Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.05340

Sigma-Aldrich

1-Chlorododecane

for synthesis

Synonym(s):

1-Chlorododecane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$47.80
1 L
$133.00

$47.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$47.80
1 L
$133.00

About This Item

Linear Formula:
CH3(CH2)11Cl
CAS Number:
Molecular Weight:
204.78
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-981-5
NACRES:
NA.22

$47.80


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥95% (GC)

form

liquid

autoignition temp.

190 °C

potency

17300 mg/kg LD50, oral (Rat)

bp

260 °C/1013 hPa

mp

-9 °C

transition temp

flash point 130 °C

solubility

<0.007 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCCCCCCCCCCCC

InChI

1S/C12H25Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3

InChI key

YAYNEUUHHLGGAH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95 % (a/a)
Density (d 20 °C/ 4 °C): 0.864 - 0.867
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service