Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.04807

Sigma-Aldrich

Isophorone

for synthesis

Synonym(s):

Isophorone, 3,5,5-Trimethyl-2-cyclohexen-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$34.90
1 L
$64.80
2.5 L
$116.00
25 L
$911.00

$34.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$34.90
1 L
$64.80
2.5 L
$116.00
25 L
$911.00

About This Item

Empirical Formula (Hill Notation):
C9H14O
CAS Number:
Molecular Weight:
138.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-126-0
NACRES:
NA.22

$34.90


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.33 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

460 °C

potency

1500 mg/kg LD50, oral (Rat)
1500 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-3.8 % (v/v)

bp

210-216 °C/1013 hPa

mp

-8 °C

transition temp

flash point 85 °C

solubility

14.5 g/L

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C1CC(CC(=C1)C)(C)C

InChI

1S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3

InChI key

HJOVHMDZYOCNQW-UHFFFAOYSA-N

Application

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.920 - 0.922
Identity (IR): passes test
Color (visual): colorless to yellowish

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service