Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.04537

Sigma-Aldrich

3-Hydroxybenzoic acid

for synthesis

Synonym(s):

3-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(OH)C6H4COOH
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-726-5

Quality Level

Assay

≥98.0% (HPLC)

pH

3 ( in H2O, Aqueous solution)

mp

199-202 °C

transition temp

flash point 220 °C

bulk density

500 kg/m3

InChI

1S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

IJFXRHURBJZNAO-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 199 °C
Melting range (upper value): ≤ 202 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service