Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03774

Sigma-Aldrich

2,4-Dichlorophenol

for synthesis

Synonym(s):

2,4-Dichlorophenol, 2,4-DCP

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$85.40

$85.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$85.40

About This Item

Linear Formula:
2,4-(Cl)2C6H3OH
CAS Number:
Molecular Weight:
163.00
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-429-6
NACRES:
NA.22

$85.40


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

potency

780 mg/kg LD50, skin (Rat)

bp

209-211 °C/1013 hPa

mp

40-43 °C

transition temp

flash point 114 °C

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)O

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

HFZWRUODUSTPEG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 41 °C
Melting range (upper value): ≤ 44 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.1 °F - closed cup

Flash Point(C)

110.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service