Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.03575

Sigma-Aldrich

2,6-Diaminopyridine

for synthesis

Synonym(s):

2,6-Diaminopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,6-(NH2)2C5H3N
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352116
EC Index Number:
205-507-2
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

solid

pH

>7 ( in H2O)

bp

159 °C/13 hPa

mp

115-118 °C

transition temp

flash point 155 °C

storage temp.

2-30°C

InChI

1S/C5H7N3/c6-4-2-1-3-5(7)8-4/h1-3H,(H4,6,7,8)

InChI key

VHNQIURBCCNWDN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 117 °C
Melting range (upper value): ≤ 121 °C
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service