Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03549

Sigma-Aldrich

Dibenzyl sulfoxide

for synthesis

Synonym(s):

Dibenzyl sulfoxide, Benzyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)SO
CAS Number:
Molecular Weight:
230.33
UNSPSC Code:
12352119
EC Index Number:
210-668-7

Assay

≥99.0% (HPLC)

pH

6.0-6.5 (20 °C, 0.3 g/L in H2O)

mp

128-131 °C

solubility

0.3 g/L

bulk density

400‑500 kg/m3

storage temp.

2-30°C

InChI

1S/C14H14OS/c15-16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

HTMQZWFSTJVJEQ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 129 °C
Melting range (upper value): ≤ 133 °C
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service