Skip to Content
MilliporeSigma
All Photos(2)

Documents

8.03057

Sigma-Aldrich

4-(Dimethylamino)benzaldehyde

for synthesis

Synonym(s):

4-(Dimethylamino)benzaldehyde, p-Formyldimethylaniline, Ehrlich′s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-[(CH3)2N]C6H4CHO
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
12352114
EC Index Number:
202-819-0
NACRES:
NA.22

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

powder

autoignition temp.

445 °C

potency

>6400 mg/kg LD50, oral (Rat)

mp

75 °C

transition temp

flash point >100 °C

density

1.22 g/cm3 at 20.1 °C

bulk density

350 kg/m3

storage temp.

no temp limit

InChI

1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3

InChI key

BGNGWHSBYQYVRX-UHFFFAOYSA-N

Application

  • Synthesis, characterization and antioxidant activity: This article explores the synthesis of Nickel (II) Schiff Base Complexes derived from 4-(dimethylamino) benzaldehyde, emphasizing their antioxidant activities (N Nawaz, I Ahmad, NM Darwesh, A Wahab, 2020).

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 70 °C
Melting range (upper value): ≤ 75 °C
Identity (IR): passes test
Related substances (TLC): ≤ 0.2 %
Appearance of solution a (colour): clear
Appearance of solution b (colour): clear

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service