Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02933

Sigma-Aldrich

Diethylene glycol dibutyl ether

for synthesis

Synonym(s):

Diethylene glycol dibutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2OCH2CH2)2O
CAS Number:
Molecular Weight:
218.33
MDL number:
UNSPSC Code:
12352112
EC Index Number:
204-001-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

268 °C

potency

3900 mg/kg LD50, oral (Rat)
3555 mg/kg LD50, skin (Rabbit)

bp

254 °C/1013 hPa

mp

-60 °C

transition temp

flash point 118 °C

solubility

3 g/L

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCCCC)CCOCCCC

InChI

1S/C12H26O3/c1-3-5-7-13-9-11-15-12-10-14-8-6-4-2/h3-12H2,1-2H3

InChI key

KZVBBTZJMSWGTK-UHFFFAOYSA-N

Application


  • Transport and Association of Ions in Lithium Battery Electrolytes Based on Glycol Ether Mixed with Halogen-Free Orthoborate Ionic Liquid: This study explores the solvent properties of glycol ethers, including diethylene glycol dibutyl ether, in enhancing the performance of lithium battery electrolytes. The findings suggest that these solvents facilitate superior ionic mobility, potentially improving battery efficiency and safety (Shah FU, et al., 2017).

  • Glymes as new solvents for lipase activation and biodiesel preparation: Diethylene glycol dibutyl ether is highlighted as an effective solvent for lipase activation, which is crucial in biodiesel production. This application demonstrates the solvent′s potential in biofuel technologies, enhancing enzyme activity and stability (Tang S, et al., 2013).

  • Extraction and carrier-facilitated transport of amino acids using synthetic non-cyclic receptors through bulk liquid membrane: This research indicates the utility of diethylene glycol dibutyl ether in the selective transport and extraction of amino acids across synthetic membranes, offering insights into its applications in biotechnological and pharmaceutical industries (Joshi P, et al., 2006).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.882 - 0.884
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service