Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02912

Supelco

Dimethyl sulfoxide for headspace gas chromatography SupraSolv®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service