Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.02712

Sigma-Aldrich

Copper(II) acetylacetonate

for synthesis

Synonym(s):

Copper(II) acetylacetonate, Bis(acetylacetonato)copper(II), Bis(2,4-pentanedionato) copper(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$103.00

$103.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$103.00

About This Item

Empirical Formula (Hill Notation):
C10H14CuO4
CAS Number:
Molecular Weight:
261.76
MDL number:
UNSPSC Code:
12352103
EC Index Number:
236-477-9
NACRES:
NA.22

$103.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.13 hPa ( 163 °C)

Quality Level

form

powder

bp

160 °C/13 hPa

mp

245 °C (decomposition)

bulk density

200‑400 kg/m3

storage temp.

2-30°C

SMILES string

[Cu+2].O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C

InChI

1S/2C5H7O2.Cu/c2*1-4(6)3-5(2)7;/h2*3H,1-2H3;/q2*-1;+2

InChI key

KMOVDRADWUSGIH-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 99.0 %
Loss on drying: ≤ 0.20 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service