Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.02527

Sigma-Aldrich

Cobalt(II) acetylacetonate

for synthesis

Synonym(s):

Cobalt(II) acetylacetonate, Bis(acetylacetonato)cobalt(II), Bis(2,4-pentanedionato) cobalt(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14CoO4
CAS Number:
Molecular Weight:
257.15
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-855-6

vapor pressure

<1 hPa ( 25 °C)

Quality Level

potency

670 mg/kg LD50, oral (Rat)

mp

170-180 °C

solubility

5.1 g/L

density

0.83 g/cm3 at 20 °C

Analysis Note

Assay (ex Co): ≥ 98.0 %
Melting range (lower value): ≥ 168 °C
Melting range (upper value): ≤ 172 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service