Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.01815

Sigma-Aldrich

Benzyl bromide

for synthesis

Synonym(s):

Benzyl bromide, α-Bromotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7Br
CAS Number:
Molecular Weight:
171.03
MDL number:
UNSPSC Code:
12352101
EC Index Number:
202-847-3

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

bp

198 °C/1013 hPa

mp

-4 °C

transition temp

flash point 87 °C

density

1.44 g/cm3 at 20 °C

InChI

1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

AGEZXYOZHKGVCM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.437 - 1.440
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service